missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-2-(methoxyimino)acetamidine, 97%, Thermo Scientific™

Product Code. 15408438
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84981-64-6

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 126.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Cyano-2-(methoxyimino)acetamidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'