missing translation for 'onlineSavingsMsg'
Learn More

2-Coumaranone, 97%

Product Code. 11428547
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428547 25 g 25g
11408547 1 g 1g
11418547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428547 Supplier Thermo Scientific Alfa Aesar Supplier No. A18617.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 553-86-6 | C8H6O2 | 134.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-86-6
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00005856
InChI Key ACZGCWSMSTYWDQ-UHFFFAOYSA-N
Synonym 2-coumaranone, benzofuran-2 3h-one, 2 3h-benzofuranone, 3h-benzofuran-2-one, benzofuran-2-one, benzofuranone, 1-benzofuran-2 3h-one, isocoumaranone, 2-coumarotioiie
PubChem CID 68382
IUPAC Name 3H-1-benzofuran-2-one
SMILES C1C2=CC=CC=C2OC1=O

Specifications

Melting Point 47°C to 52°C
Density 1.22
Boiling Point 248°C to 250°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
Beilstein 114689
Formula Weight 134.13
Percent Purity 99%
Chemical Name or Material 2-Coumaranone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.