missing translation for 'onlineSavingsMsg'
Learn More

2-Coumaranone, 97%, Thermo Scientific Chemicals

Product Code. 10256200
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-86-6
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00005856
InChI Key ACZGCWSMSTYWDQ-UHFFFAOYSA-N
Synonym 2-coumaranone, benzofuran-2 3h-one, 2 3h-benzofuranone, 3h-benzofuran-2-one, benzofuran-2-one, benzofuranone, 1-benzofuran-2 3h-one, isocoumaranone, 2-coumarotioiie
PubChem CID 68382
IUPAC Name 3H-1-benzofuran-2-one
SMILES C1C2=CC=CC=C2OC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47°C to 52°C
Boiling Point 248°C to 250°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 17, 309
Formula Weight 134.13
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Coumaranone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Coumaranone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'