missing translation for 'onlineSavingsMsg'
Learn More

2-[cis-4-(Boc-amino)cyclohexyl]ethylamine, 97%

Product Code. 15429368
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429368 1 g 1g
15418006 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429368 Supplier Thermo Scientific Alfa Aesar Supplier No. H62917.03
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 509143-12-8 | C13H26N2O2 | 242.363 g/mol

2-[cis-4-(Boc-amino)cyclohexyl]ethylamine is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine is used as a intermediate for chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 509143-12-8
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD03844601
InChI Key SSZKBGFWNFAXNC-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate
PubChem CID 24720893
IUPAC Name tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CCN

Specifications

Quantity 1 g
Formula Weight 242.35
Percent Purity 97%
Chemical Name or Material 2-[cis-4-(Boc-amino)cyclohexyl]ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.