missing translation for 'onlineSavingsMsg'
Learn More

2-(chroman-4-yl)acetic Acid, TRC

Product Code. 30365899
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365899 10 mg 10mg
30352362 5 mg -
30356466 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365899 Supplier Toronto Research Chemicals Supplier No. TRCC61173310MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5655-26-5
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
IUPAC Name 2-(3,4-dihydro-2H-chromen-4-yl)acetic acid
SMILES OC(=O)CC1CCOc2ccccc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12O3/c12-11(13)7-8-5-6-14-10-4-2-1-3-9(8)10/h1-4,8H,5-7H2,(H,12,13)
Formula Weight 192.0786
Chemical Name or Material 2-(chroman-4-yl)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.