missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorotrityl Chloride, TRC
Click to view available options
Quantity:
1 g
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1-Chloro-2-(chlorodiphenylmethyl)benzene |
| CAS | 42074-68-0 |
| Molecular Formula | C19 H14 Cl2 |
| Quantity | 1 g |
| Synonym | 1-Chloro-2-(chlorodiphenylmethyl)benzene, Clotrimazole Imp. C (EP), Benzene, 1-chloro-2-(chlorodiphenylmethyl)-, 2-Chlorophenyldiphenylchloromethane, 2-Chlorophenyldiphenylmethyl chloride, Chloro(2-chlorophenyl)diphenylmethane, Diphenyl-2-chlorophenylmethyl chloride, o-Chlorotriphenylchloromethane |
| InChI Formula | InChI=1S/C19H14Cl2/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H |
| SMILES | Clc1ccccc1C(Cl)(c2ccccc2)c3ccccc3 |
| IUPAC Name | 1-chloro-2-[chloro(diphenyl)methyl]benzene |
| Molecular Weight (g/mol) | 313.22 |
| Formula Weight | 312.0473 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction