missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g

Product Code. 11418600
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418600 2 g 2g
11438600 10 g 10g
11428600 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418600 Supplier Thermo Scientific Alfa Aesar Supplier No. 044407.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42074-68-0 | C19H14Cl2 | 313.221 g/mol

2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42074-68-0
Molecular Formula C19H14Cl2
Molecular Weight (g/mol) 313.221
MDL Number MFCD00040399
InChI Key JFLSOKIMYBSASW-UHFFFAOYSA-N
Synonym 2-chlorotrityl chloride, chloro 2-chlorophenyl methylene dibenzene, 1-chloro-2-chlorodiphenylmethyl benzene, 2-chlorophenyldiphenylmethyl chloride, 2-chlorotrityl chloride resin, 2-chlorophenyl diphenylmethyl chloride, 2-chlorotrityl resin, 2-chlorotritylchloride, chloro 2-chlorophenyl diphenylmethane, 2-chlorotrityl chloride, polymer-bound
PubChem CID 94524
IUPAC Name 1-chloro-2-[chloro(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)Cl

Specifications

Physical Form Powder
Quantity 2 g
Mesh Size 100 to 200 Mesh
Chemical Name or Material 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 1.0 to 1.4 mmol/g

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.