missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorotoluene, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Chlorotoluene |
| CAS | 95-49-8 |
| Molecular Formula | C7 H7 Cl |
| Quantity | 10 g |
| Synonym | Toluene, o-chloro- (8CI), 1-Chloro-2-methylbenzene, 1-Methyl-2-chlorobenzene, 2-Chloro-1-methylbenzene, 2-Chlorotoluene, 2-Methyl-1-chlorobenzene, 2-Methylchlorobenzene, 2-Methylphenyl chloride, 2-Tolyl chloride, NSC 8766, o-Chlorotoluene, o-Methylchlorobenzene, o-Tolyl chloride |
| InChI Formula | InChI=1S/C7H7Cl/c1-6-4-2-3-5-7(6)8/h2-5H,1H3 |
| SMILES | Cc1ccccc1Cl |
| IUPAC Name | 1-chloro-2-methylbenzene |
| Molecular Weight (g/mol) | 126.58 |
| Formula Weight | 126.0236 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction