missing translation for 'onlineSavingsMsg'
Learn More

2-chlorothiophenol, 98%, Thermo Scientific™

Product Code. 10326884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10326884 25 g Glass bottle 25g
10604565 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10326884 Supplier Thermo Scientific Acros Supplier No. 224350250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6320-03-2
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.62
MDL Number MFCD00004830
InChI Key PWOBDMNCYMQTCE-UHFFFAOYSA-N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.2750g/mL
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4SH
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.275
Synonym 4-amino-2-isopropyl-5-methylphenol hydrochloride, 6-aminothymol hydrochloride, phenol, 4-amino-5-methyl-2-1-methylethyl-, hydrochloride, 4-amino-2-isopropyl-5-methyl-phenolhydrochloride, 4-amino-2-isopropyl-5-methyl-phenol hydrochloride, 4-amino-5-methyl-2-propan-2-yl phenol hydrochloride, phenol, 4-amino-5-methyl-2-1-methylethyl-, hydrochloride 1:1, 6-aminothymol hcl, 6-aminothymolhydrochlorid, 6-aminothymol hydrovhloride
SMILES SC1=CC=CC=C1Cl
IUPAC Name 2-chlorobenzene-1-thiol
PubChem CID 80603
Formula Weight 144.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-chlorothiophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.