missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophene, 98+%, Thermo Scientific™

Product Code. 11498563
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498563 250 g 250g
11488563 50 g 50g
11478563 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498563 Supplier Thermo Scientific Alfa Aesar Supplier No. A10390.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the preparation of 5-phenylthiophene derivative and N-arylhydroxamic acids.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-43-5
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00005421
InChI Key GSFNQBFZFXUTBN-UHFFFAOYSA-N
Synonym 2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
PubChem CID 7299
IUPAC Name 2-chlorothiophene
SMILES C1=CSC(=C1)Cl

Specifications

Melting Point -72°C
Density 1.285
Boiling Point 127°C to 129°C
Flash Point 22°C (71°F)
Odor Unpleasant
Refractive Index 1.548
Quantity 250 g
UN Number UN1992
Beilstein 104652
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 118.59
Percent Purity ≥98%
Chemical Name or Material 2-Chlorothiophene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.