missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiazole, 97%

Product Code. p-7033140
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11499270

Brand: Thermo Scientific Alfa Aesar H50155.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3034-52-4 | C3H2ClNS | 119.566 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-52-4
Molecular Formula C3H2ClNS
Molecular Weight (g/mol) 119.566
MDL Number MFCD00210701
InChI Key KLEYVGWAORGTIT-UHFFFAOYSA-N
Synonym 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569
PubChem CID 76429
ChEBI CHEBI:39187
IUPAC Name 2-chloro-1,3-thiazole
SMILES C1=CSC(=N1)Cl

Specifications

Quantity 1 g
Formula Weight 119.57
Percent Purity 97%
Chemical Name or Material 2-Chlorothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.