missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiazole, 97%

Product Code. 15555236 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15555236 25 g 25g
15535236 1 g 1g
15545236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15555236 Supplier Thermo Scientific Acros Supplier No. 457260250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3034-52-4 | C3H2ClNS | 119.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-52-4
Molecular Formula C3H2ClNS
Molecular Weight (g/mol) 119.57
InChI Key KLEYVGWAORGTIT-UHFFFAOYSA-N
Synonym 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569
PubChem CID 76429
ChEBI CHEBI:39187
IUPAC Name 2-chloro-1,3-thiazole
SMILES C1=CSC(=N1)Cl

Specifications

Density 1.3780g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Specific Gravity 1.378
Formula Weight 119.57
Percent Purity 97%
Chemical Name or Material 2-Chlorothiazole
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorothiazole, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.