missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroquinoline, 98%

Product Code. 10757022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10757022 1 g Glass bottle 1g
10051691 5 g Glass bottle 5g
10086203 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10757022 Supplier Thermo Scientific Acros Supplier No. 110040010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-62-4 | C9H6ClN | 163.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-62-4
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.61
MDL Number MFCD00006741
InChI Key OFUFXTHGZWIDDB-UHFFFAOYSA-N
Synonym 2-chloro-quinoline, quinoline, 2-chloro, chloroquinoline, unii-ux7rin8gew, quinoline, chloro, ccris 3977, ux7rin8gew, chioroquinoline, 2-chloro quinoline, pubchem7580
PubChem CID 11928
IUPAC Name 2-chloroquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 34°C to 37°C
Color White to Yellow
Density 1.2300g/mL
Boiling Point 265°C to 267°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, 359
Specific Gravity 1.23
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene,petrol
Formula Weight 163.61
Percent Purity 99%
Physical Form Crystalline Powder, Crystals and/or Chunks or Fused Mass
Chemical Name or Material 2-Chloroquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloroquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.