missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine, TRC

Product Code. 30354082
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354082 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354082 Supplier Toronto Research Chemicals Supplier No. TRCC38085050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloropyrimidine
CAS 1722-12-9
Molecular Formula C4H3ClN2
Quantity 50 g
Synonym 2-Chloro-1,3-pyrimidine, NSC 43544, Pyrimidin-2-yl Chloride
InChI Formula InChI=1S/C4H3ClN2/c5-4-6-2-1-3-7-4/h1-3H
SMILES Clc1ncccn1
IUPAC Name 2-chloropyrimidine
Molecular Weight (g/mol) 114.53
Formula Weight 113.9985
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.