missing translation for 'onlineSavingsMsg'
Learn More

1-(4'-Methoxy-4-biphenylylsulfonylamino)cyclohexanecarboxylic acid, 96%, Thermo Scientific™

Product Code. 15488165
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488165 1 g 1g
15498165 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488165 Supplier Thermo Scientific Alfa Aesar Supplier No. H32698.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885269-48-7
Molecular Formula C20H23NO5S
Molecular Weight (g/mol) 389.47
MDL Number MFCD06409463
InChI Key JAKSOMBIPMPTDM-UHFFFAOYSA-N
Synonym 1-4'-methoxy-1,1'-biphenyl-4-sulfonamido cyclohexane-1-carboxylic acid, 1-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido cyclohexanecarboxylic acid, 1-4'-methoxy-4-biphenylylsulfonylamino cyclohexanecarboxylic acid, 1-4'-methoxy 1,1'-biphenyl-4-sulfonyl amino cyclohexane-1-carboxylic acid
PubChem CID 44669172
IUPAC Name 1-[[4-(4-methoxyphenyl)phenyl]sulfonylamino]cyclohexane-1-carboxylic acid
SMILES COC1=CC=C(C=C1)C1=CC=C(C=C1)S(=O)(=O)NC1(CCCCC1)C(O)=O

Specifications

Melting Point >230°C
Quantity 1 g
Formula Weight 389.47
Percent Purity 96%
Chemical Name or Material 1-(4'-Methoxy-4-biphenylylsulfonylamino)cyclohexanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.