missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Specifications
Specifications
| CAS | 458532-84-8 |
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C11H15BClNO2 |
| MDL Number | MFCD04039870 |
| Quantity | 1 g |
| Synonym | 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-4-boronic acid pinacol ester, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridin-4-ylboronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloropyridine-4-boronic acid, pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16684 |
| InChI Key | UUEQDBHKMOFLDP-UHFFFAOYSA-N |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2)Cl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction