missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyridine-3-boronic acid, 97%, Thermo Scientific™

Product Code. 10572908 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572908 1 g Glass bottle 1g
10768488 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10572908 Supplier Thermo Scientific Acros Supplier No. 430640010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 381248-04-0
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03094997
InChI Key VRDAOVQZVXYRNH-UHFFFAOYSA-N
Synonym 2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
PubChem CID 2762704
IUPAC Name (2-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=CN=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 115°C to 120°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.37
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloropyridine-3-boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloropyridine-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.