missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloropyridin-4-yl)methanamine hydrochloride, MedChemExpress

Product Code. 16424676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424676 Supplier Medchem Express Supplier No. HY101771A
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (2-Chloropyridin-4-yl)methanamine hydrochloride is a selective LOXL2 inhibitor with an IC50 of 126 nM.

(2-Chloropyridin-4-yl)methanamine hydrochloride is a selective LOXL2 inhibitor with an IC50 of 126 nM.

Specifications

Chemical Name or Material (2-Chloropyridin-4-yl)methanamine hydrochloride
CAS 916210-98-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C6H8Cl2N2
Quantity 100 mg
Solubility Information H2O : ≥ 50 mg/mL (279.25 mM) ∣DMSO : ≥ 33 mg/mL (184.31 mM)
SMILES [H]Cl.ClC1=NC=CC(CN)=C1
Molecular Weight (g/mol) 179.05
Formula Weight 179.05
Percent Purity 98.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.