missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrazine, 98%, Thermo Scientific Chemicals

Catalog Number 11479342
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
14508-49-7
C4H3ClN2
114.53
MFCD00006124
GELVZYOEQVJIRR-UHFFFAOYSA-N
chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
73277
2-chloropyrazine
ClC1=CN=CC=N1
This item is not returnable. View return policy
14508-49-7
C4H3ClN2
114.53
MFCD00006124
GELVZYOEQVJIRR-UHFFFAOYSA-N
chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
73277
2-chloropyrazine
ClC1=CN=CC=N1

2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

Solubility
Soluble in Methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Density 1.284
Boiling Point 153°C to 154°C
Flash Point 57°C (134°F)
Refractive Index 1.537
Quantity 5 g
UN Number UN1993
Beilstein 107019
Solubility Information Soluble in Methanol
Formula Weight 114.54
Percent Purity 98%
Chemical Name or Material 2-Chloropyrazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'