missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrazine, 98%, Thermo Scientific Chemicals

Product Code. 10716581
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14508-49-7
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006124
InChI Key GELVZYOEQVJIRR-UHFFFAOYSA-N
Synonym chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
PubChem CID 73277
IUPAC Name 2-chloropyrazine
SMILES ClC1=CN=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2800g/mL
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5390
Quantity 10 g
Specific Gravity 1.28
Solubility Information Solubility in water: insoluble in water
Formula Weight 114.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloropyrazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloropyrazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'