missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionyl Chloride, TRC

Product Code. 30357860
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357860 10 g 10g
30349260 50 g 50g
30350103 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357860 Supplier Toronto Research Chemicals Supplier No. TRCC38047010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7623-09-8
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.97
Synonym (±)-2-Chloropropionyl Chloride, 2-Chloro-2-methylacetyl Chloride, 2-Chloropropanoyl Chloride, alpha-Chloropropanoyl Chloride, alpha-Chloropropionic Acid Chloride, alpha-Chloropropionyl Chloride, alpha-Methylchloroacetyl Chloride
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(Cl)C(=O)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H4Cl2O/c1-2(4)3(5)6/h2H,1H3
Formula Weight 125.9639
Chemical Name or Material 2-Chloropropionyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.