missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionyl chloride, 96%

Product Code. 11445967
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445967 25 g 25g
11450318 100 g 100g
11460318 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445967 Supplier Thermo Scientific Alfa Aesar Supplier No. B24503.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7623-09-8 | C3H4Cl2O | 126.96 g/mol

2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.

Solubility
Reacts with water.

Notes
Store away from oxidizing agents, active metals, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7623-09-8
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000862
InChI Key JEQDSBVHLKBEIZ-UHFFFAOYNA-N
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
PubChem CID 111019
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(C(=O)Cl)Cl

Specifications

Density 1.308
Boiling Point 109°C to 111°C
Flash Point 31°C (88°F)
Refractive Index 1.44
Quantity 25 g
UN Number UN2920
Solubility Information Reacts with water.
Formula Weight 126.97
Percent Purity 96%
Chemical Name or Material 2-Chloropropionyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.