missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropanoic acid, TRC

Product Code. 30364168
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364168 Supplier Toronto Research Chemicals Supplier No. TRCC3810405G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material 2-Chloropropanoic acid
CAS 598-78-7
Molecular Formula C3H5ClO2
Quantity 5 g
Synonym 2-Chloropropionic acid
InChI Formula InChI=1S/C3H5ClO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)
SMILES CC(Cl)C(=O)O
IUPAC Name 2-chloropropanoic acid
Molecular Weight (g/mol) 108.52
Formula Weight 107.9978
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.