Learn More
(±)-2-chlorophenylglycine, 98%, Thermo Scientific Chemicals
£93.20 - £269.00
Chemical Identifiers
CAS | 88744-36-9 |
---|---|
Molecular Formula | C8H8ClNO2 |
Molecular Weight (g/mol) | 185.61 |
InChI Key | LMIZLNPFTRQPSF-UHFFFAOYSA-N |
Synonym | 2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh |
PubChem CID | 290729 |
IUPAC Name | 2-amino-2-(2-chlorophenyl)acetic acid |
SMILES | C1=CC=C(C(=C1)C(C(=O)O)N)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11925361
|
Thermo Scientific Acros
433340250 |
25 g |
£93.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11915361
|
Thermo Scientific Acros
433341000 |
100 g |
£269.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88744-36-9 | |
185.61 | |
2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
2-amino-2-(2-chlorophenyl)acetic acid |
C8H8ClNO2 | |
LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
290729 | |
C1=CC=C(C(=C1)C(C(=O)O)N)Cl |
Specifications
88744-36-9 | |
100.0 | |
White | |
97.5% min. (HPLC) | |
C8H8ClNO2 | |
LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
2-amino-2-(2-chlorophenyl)acetic acid | |
290729 | |
98% | |
(±)-2-chlorophenylglycine |
97.5 | |
182.9°C to 183°C | |
Authentic | |
Glass bottle | |
2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
C1=CC=C(C(=C1)C(C(=O)O)N)Cl | |
185.61 | |
185.61 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- (±)-2-Chlorophenylglycine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only