missing translation for 'onlineSavingsMsg'
Learn More

2-chlorophenylacetylene, 98%

Product Code. 10169389 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10169389 5 g 5g
10656620 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10169389 Supplier Thermo Scientific Acros Supplier No. 434750050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-31-4 | C8H5Cl | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-31-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.58
MDL Number MFCD00269951
InChI Key DGLHLIWXYSGYBI-UHFFFAOYSA-N
Synonym 2-chlorophenylacetylene, 2'-chlorophenyl acetylene, 1-chloro-2-ethynyl-benzene, benzene, 1-chloro-2-ethynyl, 1-chlor-2-ethinylbenzol, o-chlorphenylacetylen, o-chlorophenylacetylene, pubchem2550, 2-chlorophenyl acetylene, 2;-chlorophenylacetylene
PubChem CID 585995
IUPAC Name 1-chloro-2-ethynylbenzene
SMILES C#CC1=CC=CC=C1Cl

Specifications

CAS Min % 97.5
Color Colorless to Yellow
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Physical Form Liquid
Assay Percent Range 97.5% min. (GC)
Boiling Point 82°C (16.0 hPa)
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 2-chlorophenylacetylene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorophenylacetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.