missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetonitrile, 98+%

Product Code. 11434574
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434574 50 g 50g
11424574 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434574 Supplier Thermo Scientific Alfa Aesar Supplier No. A13176.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2856-63-5 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2856-63-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001898
InChI Key MRDUURPIPLIGQX-UHFFFAOYSA-N
Synonym 2-chlorobenzyl cyanide, 2-chlorophenylacetonitrile, 2-2-chlorophenyl acetonitrile, o-chlorobenzyl cyanide, benzeneacetonitrile, 2-chloro, 2-chlorobenzeneacetonitrile, 2-chlorobenzylcyanide, 2-chlorophenyl acetonitrile, o-chlorophenyl acetonitrile, acetonitrile, o-chlorophenyl
PubChem CID 76112
IUPAC Name 2-(2-chlorophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)Cl

Specifications

Melting Point 21°C to 24°C
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 50 g
UN Number UN3439
Beilstein 2043683
Formula Weight 151.6
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.