missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic Acid, TRC

Product Code. 30363309
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363309 5 g -
30354536 50 g 50g
30351860 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363309 Supplier Toronto Research Chemicals Supplier No. TRCC3753805G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-36-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
Synonym (2-Chlorophenyl)acetic Acid, (o-Chlorophenyl)acetic Acid, 2-(o-Chlorophenyl)ethanoic Acid, 2-Chlorobenzeneacetic Acid, NSC 4613, o-Chlorobenzeneacetic Acid
IUPAC Name 2-(2-chlorophenyl)acetic acid
SMILES OC(=O)Cc1ccccc1Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H7ClO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)
Formula Weight 170.0135
Chemical Name or Material 2-Chlorophenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.