missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenylacetic acid, 98.5+%, Thermo Scientific Chemicals
£15.50 - £106.00
Chemical Identifiers
CAS | 2444-36-2 |
---|---|
Molecular Formula | C8H7ClO2 |
Molecular Weight (g/mol) | 170.6 |
MDL Number | MFCD00004317 |
InChI Key | IUJAAIZKRJJZGQ-UHFFFAOYSA-N |
Synonym | 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid |
PubChem CID | 17124 |
IUPAC Name | 2-(2-chlorophenyl)acetic acid |
SMILES | C1=CC=C(C(=C1)CC(=O)O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10327700
|
Thermo Scientific Acros
184030050 |
5 g | Glass bottle | ||||||
10514131
|
Thermo Scientific Acros
184030250 |
25 g | Glass bottle | ||||||
10168123
|
Thermo Scientific Acros
184031000 |
100 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2444-36-2 | |
170.6 | |
IUJAAIZKRJJZGQ-UHFFFAOYSA-N | |
17124 | |
C1=CC=C(C(=C1)CC(=O)O)Cl |
C8H7ClO2 | |
MFCD00004317 | |
2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid | |
2-(2-chlorophenyl)acetic acid |
Specifications
94.0°C to 97.0°C | |
98.5 | |
100.0 | |
Powder | |
C8H7ClO2 | |
MFCD00004317 | |
2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid | |
IUJAAIZKRJJZGQ-UHFFFAOYSA-N | |
2-(2-chlorophenyl)acetic acid | |
17124 | |
98.5+% |
2444-36-2 | |
White to Yellow | |
Authentic | |
98.5+% | |
ClC6H4CH2CO2H | |
09, 447 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol | |
C1=CC=C(C(=C1)CC(=O)O)Cl | |
170.6 | |
170.6 | |
2-Chlorophenylacetic acid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 219-482-
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Chlorophenylacetic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only