missing translation for 'onlineSavingsMsg'
Learn More

(2-Chlorophenyl)trichlormethane, Mikromol™

Product Code. 17983776
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize':
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
17983776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 17983776 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM0015.08
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2-Chlorophenyl)trichlormethane
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha,alpha,alpha-2-Tetrachlorotoluene
CAS 2136-89-2
Analyte or Component Names alpha,alpha,alpha-2-Tetrachlorotoluene
Shipping Condition Room Temperature
Molecular Formula C7 H4 Cl4
Certifications/Compliance ISO 17025
Synonym (2-Chlorophenyl)trichlormethane
InChI Formula InChI=1S/C7H4Cl4/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H
SMILES Clc1ccccc1C(Cl)(Cl)Cl
IUPAC Name 1-chloro-2-(trichloromethyl)benzene
Molecular Weight (g/mol) 229.92
Quantity 100 mg
Formula Weight 227.9067 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.