missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl phosphorodichloridate, 98+%

Product Code. 11400433
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400433 100 g 100g
11480423 5 g 5g
11490423 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400433 Supplier Thermo Scientific Alfa Aesar Supplier No. A11505.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15074-54-1 | C6H4Cl3O2P | 245.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15074-54-1
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009704
InChI Key VLDPXPPHXDGHEW-UHFFFAOYSA-N
Synonym 2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
PubChem CID 84775
IUPAC Name 1-chloro-2-dichlorophosphoryloxybenzene
SMILES C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl

Specifications

Density 1.491
Boiling Point 82°C to 84°C (0.4 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.538
Quantity 100 g
UN Number UN3265
Beilstein 911009
Sensitivity Moisture sensitive
Formula Weight 245.43
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenyl phosphorodichloridate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.