missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl isothiocyanate, 99%, Thermo Scientific Chemicals

Catalog Number 10140122
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2740-81-0
C7H4ClNS
169.63
MFCD00004801
DASSPOJBUMBXLU-UHFFFAOYSA-N
2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
123171
1-chloro-2-isothiocyanatobenzene
C1=CC=C(C(=C1)N=C=S)Cl
This item is not returnable. View return policy
2740-81-0
C7H4ClNS
169.63
MFCD00004801
DASSPOJBUMBXLU-UHFFFAOYSA-N
2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
123171
1-chloro-2-isothiocyanatobenzene
C1=CC=C(C(=C1)N=C=S)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
Color Yellow
CAS Max % 100.0
Density 1.2900g/mL
Boiling Point 260°C to 262°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.6598 to 1.6618
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.29
Linear Formula NCSC6H4Cl
Beilstein 12, 601
Formula Weight 169.63
Percent Purity 99%
Chemical Name or Material 2-Chlorophenyl isothiocyanate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorophenyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'