missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl cyclopentyl ketone, 97%

Product Code. 11332526
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332526 50 g 50g
11322526 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332526 Supplier Thermo Scientific Alfa Aesar Supplier No. L06448.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6740-85-8 | C12H13ClO | 208.685 g/mol

2-Chlorophenyl cyclopentyl ketone is used as a reagent in the preparation of bicyclic and polycyclic aromatic hydrocarbons. It acts as a pharmaceutical intermediate and also as an intermediate of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorophenyl cyclopentyl ketone is used as a reagent in the preparation of bicyclic and polycyclic aromatic hydrocarbons. It acts as a pharmaceutical intermediate and also as an intermediate of ketamine.

Solubility
Slightly miscible with chloroform and ethyl acetate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6740-85-8
Molecular Formula C12H13ClO
Molecular Weight (g/mol) 208.685
MDL Number MFCD00038367
InChI Key QIJMMRNZBJHXRI-UHFFFAOYSA-N
Synonym 2-chlorophenyl cyclopentyl ketone, 2-chlorophenyl cyclopentyl methanone, o-chlorophenyl cyclopentyl ketone, o-chlorobenzoylcyclopentane, 2-chlorophenyl-cyclopentylketone, methanone, 2-chlorophenyl cyclopentyl, 2-chlorobenzoylcyclopentane, acmc-1b5a4, 0-chlorphenylcyclopentylketone, 1-2-chlorobenzoyl cyclopentane
PubChem CID 81223
IUPAC Name (2-chlorophenyl)-cyclopentylmethanone
SMILES C1CCC(C1)C(=O)C2=CC=CC=C2Cl

Specifications

Density 1.16
Boiling Point 130°C (5 mmHg)
Flash Point >100°C (212°F)
Odor Aromatic
Refractive Index 1.547
Quantity 50 g
Beilstein 1873022
Solubility Information Slightly miscible with chloroform and ethyl acetate.
Formula Weight 208.69
Percent Purity 97%
Chemical Name or Material 2-Chlorophenyl cyclopentyl ketone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.