missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenoxyacetonitrile, 97%, Thermo Scientific™

Product Code. 11493764
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493764 100 g 100g
11473764 5 g 5g
11483764 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493764 Supplier Thermo Scientific Alfa Aesar Supplier No. A11346.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43111-31-5
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00017331
InChI Key JWRJGSQLKHGQOT-UHFFFAOYSA-N
Synonym 2-2-chlorophenoxy acetonitrile, 2-chlorophenoxyacetonitrile, 2-chlorophenoxy-acetonitrile, 2-2-chlorophenoxy ethanenitrile, acmc-20anxp, o-chlorophenoxy acetonitrile, 2-chloro-phenoxy-acetonitrile, 2-2-chloranylphenoxy ethanenitrile, acetonitrile,2-2-chlorophenoxy
PubChem CID 521134
IUPAC Name 2-(2-chlorophenoxy)acetonitrile
SMILES C1=CC=C(C(=C1)OCC#N)Cl

Specifications

Quantity 100 g
Refractive Index 1.542
UN Number UN3276
Beilstein 2089865
Boiling Point 109°C to 110°C (1 mmHg)
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 2-Chlorophenoxyacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.