missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenoxyacetic acid, 98+%

Product Code. 11324374
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324374 100 g 100g
11314374 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324374 Supplier Thermo Scientific Alfa Aesar Supplier No. L00926.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 614-61-9 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-61-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00004298
InChI Key OPQYFNRLWBWCST-UHFFFAOYSA-N
Synonym 2-chlorophenoxyacetic acid, 2-chlorophenoxy acetic acid, o-chlorophenoxyacetic acid, 2-2-chlorophenoxy acetic acid, acetic acid, 2-chlorophenoxy, acide o-chlorophenoxyacetique, acetic acid, o-chlorophenoxy, unii-x2512m5l7q, acetic acid, chlorophenoxy
PubChem CID 11969
IUPAC Name 2-(2-chlorophenoxy)acetic acid
SMILES OC(=O)COC1=CC=CC=C1Cl

Specifications

Melting Point 147°C to 152°C
Quantity 100 g
Beilstein 1103151
Formula Weight 186.59
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.