missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenol, 98+%

Product Code. 10239603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Can
Unit Size:
100g
10kg
2.5kg
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239603 2.5 kg Glass bottle 2.5kg
10722231 5 g Glass bottle 5g
10072250 100 g Glass bottle 100g
10635632 500 g Glass bottle 500g
11954681 10 kg Plastic Can 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10239603 Supplier Thermo Scientific Acros Supplier No. 180990025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-57-8 | C6H5ClO | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-57-8
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002159
InChI Key ISPYQTSUDJAMAB-UHFFFAOYSA-N
Synonym o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
PubChem CID 7245
ChEBI CHEBI:47083
IUPAC Name 2-chlorophenol
SMILES C1=CC=C(C(=C1)O)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 8.0°C
Density 1.2620g/mL
Boiling Point 173.0°C to 175.0°C
Flash Point 63°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4OH
Refractive Index 1.5580 to 1.5600
Quantity 2.5 kg
Beilstein 06,183
Merck Index 15,2157
Specific Gravity 1.262
Solubility Information Solubility in water: 28.5g/l (20°C). Other solubilities: freely soluble in solutions of sodium hydroxide,, alcohol and ether, soluble in aromatic solvents
Viscosity 4.11 mPa.s (25°C)
Formula Weight 128.56
Percent Purity 98+%
Chemical Name or Material 2-Chlorophenol, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.