missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenethylalcohol, 98%, Thermo Scientific Chemicals
£17.10 - £93.90
Chemical Identifiers
CAS | 19819-95-5 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.61 |
MDL Number | MFCD00002888 |
InChI Key | IWNHTCBFRSCBQK-UHFFFAOYSA-N |
Synonym | 2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol |
PubChem CID | 88266 |
IUPAC Name | 2-(2-chlorophenyl)ethanol |
SMILES | C1=CC=C(C(=C1)CCO)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10484205
|
Thermo Scientific Acros
186190010 |
1 g | Glass bottle |
£17.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10393522
|
Thermo Scientific Acros
186190050 |
5 g | Glass bottle |
£28.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10038511
|
Thermo Scientific Acros
186190250 |
25 g | Glass bottle |
£93.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19819-95-5 | |
156.61 | |
IWNHTCBFRSCBQK-UHFFFAOYSA-N | |
88266 | |
C1=CC=C(C(=C1)CCO)Cl |
C8H9ClO | |
MFCD00002888 | |
2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol | |
2-(2-chlorophenyl)ethanol |
Specifications
19819-95-5 | |
Colorless | |
1.1900g/mL | |
Authentic | |
1.5495 to 1.5515 | |
1.19 | |
ClC6H4CH2CH2OH | |
2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol | |
C1=CC=C(C(=C1)CCO)Cl | |
156.61 | |
156.61 | |
2-Chlorophenethylalcohol, 98% |
97.5 | |
100.0 | |
84.0°C to 85.0°C (3.0 mmHg) | |
Liquid | |
97.5% min. (GC) | |
C8H9ClO | |
MFCD00002888 | |
IWNHTCBFRSCBQK-UHFFFAOYSA-N | |
2-(2-chlorophenyl)ethanol | |
88266 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 243-348-0
missing translation for 'tsca' : TSCA
RUO – Research Use Only