missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenethylalcohol, 98%

Product Code. 10038511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10038511 25 g Glass bottle 25g
10484205 1 g Glass bottle 1g
10393522 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10038511 Supplier Thermo Scientific Acros Supplier No. 186190250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19819-95-5 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19819-95-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00002888
InChI Key IWNHTCBFRSCBQK-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol
PubChem CID 88266
IUPAC Name 2-(2-chlorophenyl)ethanol
SMILES C1=CC=C(C(=C1)CCO)Cl

Specifications

CAS Min % 97.5
Color Colorless
CAS Max % 100.0
Density 1.1900g/mL
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5495 to 1.5515
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.19
Linear Formula ClC6H4CH2CH2OH
Boiling Point 84.0°C to 85.0°C (3.0 mmHg)
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 2-Chlorophenethylalcohol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.