missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinic acid, 99%

Product Code. 10209790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 10209790

Brand: Thermo Scientific Acros 404860500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2942-59-8 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2942-59-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 176°C to 178°C
Color White to Cream
Loss on Drying 0.3% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H4ClNO2
Linear Formula Cl(C5H3N)COOH
MDL Number MFCD00006236
Quantity 50 g
Beilstein 22, II, 35
Synonym 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
InChI Key IBRSSZOHCGUTHI-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)Cl)C(=O)O
IUPAC Name 2-chloropyridine-3-carboxylic acid
Molecular Weight (g/mol) 157.56
PubChem CID 76258
Formula Weight 157.56
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Chloronicotinic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloronicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.