missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinic acid, 99%

Product Code. 11468956
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468956 250 g 250g
11448956 10 g 10g
11458956 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468956 Supplier Thermo Scientific Alfa Aesar Supplier No. A17082.30
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2942-59-8 | C6H4ClNO2 | 157.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-59-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00006236
InChI Key IBRSSZOHCGUTHI-UHFFFAOYSA-N
Synonym 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
PubChem CID 76258
IUPAC Name 2-chloropyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Cl)C(=O)O

Specifications

Melting Point ∼180°C (decomposition)
Quantity 250 g
Beilstein 119023
Formula Weight 157.56
Percent Purity 99%
Chemical Name or Material 2-Chloronicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.