missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)pyridine hydrochloride, 98%

Product Code. 11403923
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403923 100 g 100g
11483913 5 g 5g
11493913 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403923 Supplier Thermo Scientific Alfa Aesar Supplier No. A10226.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6959-47-3 | C6H7Cl2N | 164.029 g/mol

2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.

Solubility
Soluble in water, acetone (10-50mg/ml), 95%ethanol (100mg/ml).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6959-47-3
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00012811
InChI Key JPMRGPPMXHGKRO-UHFFFAOYSA-N
Synonym 2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
PubChem CID 23392
ChEBI CHEBI:76600
IUPAC Name 2-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC=NC(=C1)CCl.Cl

Specifications

Melting Point 120°C to 124°C
Quantity 100 g
UN Number UN3261
Beilstein 3689155
Sensitivity Hygroscopic
Solubility Information Soluble in water,acetone (10-50mg/ml),95%ethanol (100mg/ml).
Formula Weight 164.04
Percent Purity 98%
Chemical Name or Material 2- (Chloromethyl)pyridine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.