missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)benzimidazole, 95%

Product Code. 11497226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497226 25 g 25g
11487226 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497226 Supplier Thermo Scientific Alfa Aesar Supplier No. B25548.14
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4857-04-9 | C8H7ClN2 | 166.61 g/mol

2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-04-9

Specifications

Melting Point 141°C to 143°C (decomposition)
Molecular Formula C7H5ClN2
MDL Number MFCD00051944
Quantity 25 g
Synonym 2-chlorobenzimidazole, 2-chloro-1h-benzo d imidazole, 1h-benzimidazole, 2-chloro, 2-chloro-1h-benzoimidazole, 2-chloro-1h-1,3-benzodiazole, benzimidazole, 2-chloro, 2-chloro-1h-1,3-benzimidazole, 2-chlorobenzoimidazole, pubchem7598, 2-chloro benzimidazole
Solubility Information Insoluble in water.
InChI Key AYPSHJCKSDNETA-UHFFFAOYSA-N
SMILES ClC1=NC2=CC=CC=C2N1
IUPAC Name 2-chloro-1H-benzimidazole
Molecular Weight (g/mol) 152.58
PubChem CID 78572
Formula Weight 166.61
Percent Purity 96%
Chemical Name or Material 2-(Chloromethyl)benzimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.