missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloromethyl-3-methyl-4-(2,2,2-trifluoro-ethoxy)pyridine Hydrochloride, Mikromol™
Description
2-Chloromethyl-3-methyl-4-(2,2,2-trifluoro-ethoxy)pyridine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 2-Chloromethyl-3-methyl-4-(2,2,2-trifluoro-ethoxy)pyridine Hydrochloride |
| CAS | 127337-60-4 |
| Analyte or Component Names | 2-Chloromethyl-3-methyl-4-(2,2,2-trifluoro-ethoxy)pyridine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H9 Cl F3 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Chloromethyl-3-methyl-4-(2,2,2-trifluoro-ethoxy)pyridine Hydrochloride |
| InChI Formula | InChI=1S/C9H9ClF3NO.ClH/c1-6-7(4-10)14-3-2-8(6)15-5-9(11,12)13;/h2-3H,4-5H2,1H3;1H |
| SMILES | Cl.Cc1c(CCl)nccc1OCC(F)(F)F |
| IUPAC Name | 2-(chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?