missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-2-phenylpropane, TRC

Product Code. 30359658
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30359658

Brand: Toronto Research Chemicals TRCC3680801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloromethyl-2-phenylpropane
CAS 515-40-2
Molecular Formula C10H13Cl
Quantity 1 g
Synonym (1,1-Dimethyl-2-chloroethyl)benzene, (beta-Chloro-tert-butyl)benzene, (beta-Chloro-alpha,alpha-dimethyl)ethylbenzene, 1-Chloro-2-methyl-2-phenylpropane, 2-Methyl-2-phenylpropyl Chloride, NSC 54159, NSC 923, Neophyl Chloride, beta,beta-Dimethylphenethyl Chloride
InChI Formula InChI=1S/C10H13Cl/c1-10(2,8-11)9-6-4-3-5-7-9/h3-7H,8H2,1-2H3
SMILES CC(C)(CCl)c1ccccc1
IUPAC Name (1-chloro-2-methylpropan-2-yl)benzene
Molecular Weight (g/mol) 168.66
Formula Weight 168.0706
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.