missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromandelic acid, TRC

Product Code. 30368437
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368437 50 g 50g
30355542 25 g 25g
30362133 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368437 Supplier Toronto Research Chemicals Supplier No. TRCC43267350G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10421-85-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
IUPAC Name 2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES OC(C(=O)O)c1ccccc1Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H7ClO3/c9-6-4-2-1-3-5(6)7(10)8(11)12/h1-4,7,10H,(H,11,12)
Formula Weight 186.0084
Chemical Name or Material 2-Chloromandelic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.