missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromandelic acid, 98%

Product Code. 11497084
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497084 25 g 25g
11487084 5 g 5g
11407094 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497084 Supplier Thermo Scientific Alfa Aesar Supplier No. B21525.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 10421-85-9 | C8H7ClO3 | 186.59 g/mol

2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.

Solubility
Very soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10421-85-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00084962
InChI Key RWOLDZZTBNYTMS-UHFFFAOYNA-N
Synonym 2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895
PubChem CID 97720
IUPAC Name 2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=CC=C1Cl

Specifications

Melting Point 80°C to 85°C
Quantity 25 g
Beilstein 2367070
Solubility Information Very soluble in water.
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 2-Chloromandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.