missing translation for 'onlineSavingsMsg'
Learn More

2-chloromalonaldehyde, 95%

Product Code. 10697943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697943 50 g Glass bottle 50g
10013810 1 g Glass bottle 1g
10396441 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10697943 Supplier Thermo Scientific Acros Supplier No. 330120500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36437-19-1 | C3H3ClO2 | 106.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36437-19-1
MDL Number MFCD01320489
InChI Key KTRZQCIGJUWSGE-UHFFFAOYSA-N
Synonym 2-chloromalonaldehyde, chloromalonaldehyde, propanedial, chloro, chloromalonalde-hyde, 2-chloro-malonaldehyde, propanedial, 2-chloro, 2-chloropropane-1,3-dial, chloromalonaldehyde; chloromalondialdehyde; chloromalonic dialdehyde;2-chloromalonaldehyde
PubChem CID 147646
IUPAC Name 2-chloropropanedial
SMILES C(=O)C(C=O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 140.0°C to 145.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C3H3ClO2
Linear Formula ClCH(CHO)2
Quantity 50 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,benzene,dimethylsulfoxide
Molecular Weight (g/mol) 106.5
Formula Weight 106.5
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloromalonaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.