Learn More
alpha-2-Chloroisodurene, 98%, Thermo Scientific Chemicals
£15.40 - £43.30
Chemical Identifiers
CAS | 1585-16-6 |
---|---|
Molecular Formula | C10H13Cl |
Molecular Weight (g/mol) | 168.66 |
MDL Number | MFCD00000901 |
InChI Key | UNRGEIXQCZHICP-UHFFFAOYSA-N |
Synonym | 2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene |
PubChem CID | 74108 |
IUPAC Name | 2-(chloromethyl)-1,3,5-trimethylbenzene |
SMILES | CC1=CC(C)=C(CCl)C(C)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10062030
|
Thermo Scientific Acros
159780050 |
5 g | Glass bottle |
£15.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10467000
|
Thermo Scientific Acros
159780250 |
25 g | Glass bottle |
£43.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1585-16-6 | |
168.66 | |
UNRGEIXQCZHICP-UHFFFAOYSA-N | |
74108 | |
CC1=CC(C)=C(CCl)C(C)=C1 |
C10H13Cl | |
MFCD00000901 | |
2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene | |
2-(chloromethyl)-1,3,5-trimethylbenzene |
Specifications
1585-16-6 | |
100.0 | |
White | |
106°C | |
97.5% min. (GC) | |
(CH3)3C6H2CH2Cl | |
2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene | |
UNRGEIXQCZHICP-UHFFFAOYSA-N | |
2-(chloromethyl)-1,3,5-trimethylbenzene | |
74108 | |
98% | |
α-2-Chloroisodurene, 98% |
97.5 | |
34°C to 38°C | |
114°C to 115°C (10.0 mmHg) | |
Authentic | |
C10H13Cl | |
MFCD00000901 | |
Solubility in water: insoluble | |
CC1=CC(C)=C(CCl)C(C)=C1 | |
168.66 | |
168.67 | |
Fused Solid, Crystals, or Chunks |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-440-3
missing translation for 'tsca' : TSCA
- alpha-2-Chloroisodurene
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only