missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl vinyl ether, ca. 95%

Product Code. 10306890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10306890 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10306890 Supplier Thermo Scientific Acros Supplier No. 149431000
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-75-8 | C4H7ClO | 106.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-75-8
CAS Min % 94.0
CAS Max % 100.0
Melting Point -70°C
Color Colorless to Yellow
Density 1.0480g/mL
Boiling Point 109°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7ClO
Linear Formula ClCH2CH2OCH=CH2
Refractive Index 1.4370 to 1.439
MDL Number MFCD00000973
Quantity 100 g
Beilstein 01, II, 473
Merck Index 15, 2140
Specific Gravity 1.048
Synonym 2-chloroethyl vinyl ether, 2-chloroethoxy ethene, ethene, 2-chloroethoxy, 2-vinyloxyethyl chloride, vinyl 2-chloroethyl ether, ether, 2-chloroethyl vinyl, rcra waste number u042, beta-chloroethyl vinyl ether, 2-chlorethyl vinyl ether
Solubility Information Solubility in water: slightly soluble. Other solubilities: hydrolysis in alkaline waters
InChI Key DNJRKFKAFWSXSE-UHFFFAOYSA-N
SMILES C=COCCCl
IUPAC Name 1-chloro-2-ethenoxyethane
Molecular Weight (g/mol) 106.55
PubChem CID 8074
Formula Weight 106.55
Percent Purity ca. 95%
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl vinyl ether, ca. 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.