missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.)

Product Code. 11487517
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487517 5 g 5g
11497517 25 g 25g
11407527 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487517 Supplier Thermo Scientific Alfa Aesar Supplier No. A15630.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 999-81-5 | C5H13Cl2N | 158.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-81-5
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00011869
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
Synonym chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
PubChem CID 13836
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
SMILES C[N+](C)(C)CCCl.[Cl-]

Specifications

Melting Point ∼241°C (decomposition)
Odor Amine-like
Linear Formula ClCH2CH2N(CH3)3Cl
Quantity 5 g
UN Number UN2811
Beilstein 3563994
Sensitivity Very Hygroscopic
Merck Index 14,2104
Formula Weight 158.07
Percent Purity 98%
Assay (dry wt.), may cont. up to 5% water
Chemical Name or Material (2-Chloroethyl)trimethylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.