missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)trimethylammonium chloride, 98%

Product Code. 10246890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10246890 100 g 100g
10052680 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10246890 Supplier Thermo Scientific Acros Supplier No. 215421000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 999-81-5 | C5H13Cl2N | 158.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-81-5
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.07
MDL Number MFCD00011869
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
Synonym chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
PubChem CID 13836
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
SMILES C[N+](C)(C)CCCl.[Cl-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 239°C to 243°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCH2CH2N(CH3)3Cl
Quantity 100 g
Beilstein 04, 134
Merck Index 15, 2104
Solubility Information Solubility in water: 740g/L (20°C)
Formula Weight 158.07
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (2-Chloroethyl)trimethylammonium chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.